3-Chlorosalicylic acid ≥95%

Supplier: Matrix Scientific

076333-5G 076333-25G
102545-626EA 187.69 USD
102545-626 102833-918
3-Chlorosalicylic acid ≥95%
3-Chlorosalicylic acid

Hazard: Irritant

Formula: C₇H₅ClO₃
MW: 172.57 g/mol
Melting Pt: 184…186 °C
MDL Number: MFCD00010390
CAS Number: 1829-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR