1H-Pyrrolo[2,3-b]pyridin-4-amine acetate ≥95%

Supplier: Matrix Scientific

076331-250MG 076331-1G 076331-5G
102545-612EA 400 USD
102545-612 102545-614 102545-616
1H-Pyrrolo[2,3-b]pyridin-4-amine acetate ≥95%
1H-Pyrrolo[2,3-b]pyridin-4-amine acetate

Hazard: Irritant

Formula: C₉H₁₁N₃O₂
CAS Number: 1373253-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR