tert-Butyl-2,2-dimethylpiperazine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 2,2-dimethylpiperazine-1-carboxylate
076327-250MG
076327-1G
102545-596EA
958.46
USD
102545-596
102545-598
tert-Butyl-2,2-dimethylpiperazine-1-carboxylate ≥95%
tert-Butyl-2,2-dimethylpiperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₁H₂₂N₂O₂ MW: 214.31 g/mol |
MDL Number:
MFCD07371652 CAS Number: 674792-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...