3-Methyl-5-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

076325-1G 076325-5G
102545-586EA 533.85 USD
102545-586 102545-588
3-Methyl-5-nitrobenzoic acid ≥95%
3-Methyl-5-nitrobenzoic acid

Hazard: Irritant

Formula: C₈H₇NO₄
MDL Number: MFCD00129754
CAS Number: 113882-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR