4-Methoxyphenylboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Methoxyphenylboronic acid Pinacol ester

076322-1G 076322-5G
102545-574EA 58.46 USD
102545-574 102545-576
4-Methoxyphenylboronic acid pinacol ester ≥95%
4-Methoxyphenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₃H₁₉BO₃
MW: 234.1 g/mol
MDL Number: MFCD05663864
CAS Number: 171364-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR