4-Nitro-2,3-xylidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-Dimethyl-4-nitroaniline

076321-1G 076321-5G
102545-570EA 533.85 USD
102545-570 102545-572
4-Nitro-2,3-xylidine ≥95%
4-Nitro-2,3-xylidine

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.1772 g/mol
MDL Number: MFCD00130024
CAS Number: 80879-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR