2-Chloro-3-methoxyquinoxaline ≥95%

Supplier: Matrix Scientific

076318-250MG 076318-1G
102545-556EA 1110.77 USD
102545-556 102545-558
2-Chloro-3-methoxyquinoxaline ≥95%
2-Chloro-3-methoxyquinoxaline

Hazard: Irritant

Formula: C₉H₇ClN₂O
MW: 194.6200 g/mol
MDL Number: MFCD07367979
CAS Number: 32998-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR