Methyl-2-bromobenzo[d]thiazole-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-bromobenzo[d]thiazole-6-carboxylate

076317-250MG 076317-1G 076317-5G
102545-550EA 355.38 USD
102545-550 102545-552 102545-554
Methyl-2-bromobenzo[d]thiazole-6-carboxylate ≥95%
Methyl-2-bromobenzo[d]thiazole-6-carboxylate

Hazard: Irritant

Formula: C₉H₆BrNO₂S
MDL Number: MFCD16660646
CAS Number: 1024583-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR