Boc-8-amino-3,6-dioxaoctanoic acid DCHA ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-AEEA.DCHA, Dicyclohexylamine 2,2-dimethyl-4-oxo-3,8,11-trioxa-5-azatridecan-13-oate

076313-250MG 076313-1G 076313-5G
102545-530EA 284.62 USD
102545-530 102545-532 102545-534
Boc-8-amino-3,6-dioxaoctanoic acid DCHA ≥95%
Boc-8-amino-3,6-dioxaoctanoic acid DCHA

Hazard: Irritant

Formula: C₂₃H₄₄N₂O₆
MW: 444.62 g/mol
MDL Number: MFCD06796877
CAS Number: 560088-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR