N-(3-Hydroxypropyl)morpholine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Morpholinopropanol, 3-Morpholino-1-propanol, 4-(3-Hydroxypropyl)morpholine, 3-(4-Morpholinyl)-1-propanol

076312-1G 076312-5G 076312-10G
102545-524EA 72.31 USD
102545-524 102545-526 102545-528
N-(3-Hydroxypropyl)morpholine ≥95%
N-(3-Hydroxypropyl)morpholine

Hazard: Irritant

Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
MDL Number: MFCD01713533
CAS Number: 4441-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR