5-Bromo-2-chloro-3-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

076311-1G 076311-5G
102545-518EA 30.77 USD
102545-518 102545-520
5-Bromo-2-chloro-3-(trifluoromethyl)pyridine ≥95%
5-Bromo-2-chloro-3-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂BrClF₃N
MW: 260.44 g/mol
MDL Number: MFCD11100983
CAS Number: 211122-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR