Ethyl-2-methylthiophene-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-methylthiophene-3-carboxylate

076310-250MG 076310-1G 076310-5G
102545-512EA 355.38 USD
102545-512 102545-514 102545-516
Ethyl-2-methylthiophene-3-carboxylate ≥95%
Ethyl-2-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₈H₁₀O₂S
MW: 170.2300 g/mol
MDL Number: MFCD14698267
CAS Number: 19432-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR