3-Iodobenzylamine hydrochloride ≥95%
Supplier: Matrix Scientific
076308-1G
076308-5G
102545-504EA
78.46
USD
102545-504
102545-506
3-Iodobenzylamine hydrochloride ≥95%
3-Iodobenzylamine hydrochloride
Hazard: Irritant
Formula:
C₇H₉ClIN |
MDL Number:
MFCD00012857 CAS Number: 3718-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...