Cyclopentanone-2-carbonitrile ≥95%

Supplier: Matrix Scientific

076305-1G 076305-5G 076305-10G
102545-486EA 61.54 USD
102545-486 102545-488 102545-490
Cyclopentanone-2-carbonitrile ≥95%
Cyclopentanone-2-carbonitrile

Hazard: Irritant-Harmful/Refrigerate

Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00154838
CAS Number: 2941-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR