cis-4-Hydroxy-L-proline methyl ester hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: H-cis-Hyp-OMe·HCl, Methyl cis-4-Hydroxy-L-proline Hydrochloride

076301-1G 076301-5G
102545-466EA 81.54 USD
102545-466 102545-468
cis-4-Hydroxy-L-proline methyl ester hydrochloride ≥95%
cis-4-Hydroxy-L-proline methyl ester hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO₃
MDL Number: MFCD00237835
CAS Number: 40126-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR