3-Bromo-4-methyl-5-nitropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-4-methy-5nitro-pyridine

076300-1G 076300-5G
102545-460EA 190.77 USD
102545-460 102545-462
3-Bromo-4-methyl-5-nitropyridine ≥95%
3-Bromo-4-methyl-5-nitropyridine

Hazard: Irritant

Formula: C6H5BrN2O2
MW: 217.0200 g/mol
MDL Number: MFCD11847767
CAS Number: 69872-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR