1H-Pyrrolo[3,2-c]pyridine-3-carboxaldehyde ≥95%

Supplier: Matrix Scientific

076299-1G 076299-5G
102545-456EA 353.85 USD
102545-456 102545-458
1H-Pyrrolo[3,2-c]pyridine-3-carboxaldehyde ≥95%
1H-Pyrrolo[3,2-c]pyridine-3-carboxaldehyde

Hazard: Irritant

Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD08690128
CAS Number: 933717-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR