1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene ≥95%

Supplier: Matrix Scientific

076298-25G 076298-100G
102545-450EA 98.46 USD
102545-450 102545-452
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene ≥95%
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene

Hazard: Irritant

Formula: C₇H₃BrF₄O
MDL Number: MFCD01861132
CAS Number: 105529-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR