2-Thiazoleacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Thiazolyl)acetic Acid

076296-250MG 076296-1G
102545-442EA 541.54 USD
102545-442 102545-444
2-Thiazoleacetic acid ≥95%
2-Thiazoleacetic acid

Hazard: Irritant

Formula: C₅H₅NO₂S
MW: 143.17 g/mol
MDL Number: MFCD02094168
CAS Number: 188937-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR