2,4,6-Trichlorobenzyl alcohol ≥95%

Supplier: Matrix Scientific

076292-1G 076292-5G
102545-426EA 178.46 USD
102545-426 102545-428
2,4,6-Trichlorobenzyl alcohol ≥95%
2,4,6-Trichlorobenzyl alcohol

Hazard: Irritant

Formula: C₇H₅Cl₃O
MDL Number: MFCD03093891
CAS Number: 217479-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR