Ethyl-4-boc-2-morpholinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 4-Boc-2-morpholinecarboxylate
076291-1G
076291-5G
102545-422EA
569.23
USD
102545-422
102545-424
Ethyl-4-boc-2-morpholinecarboxylate ≥95%
Ethyl-4-boc-2-morpholinecarboxylate
Hazard: Irritant
Formula:
C₁₂H₂₁NO₅ MW: 259.2988 g/mol |
MDL Number:
MFCD01646390 CAS Number: 768371-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...