EMCS (6-Maleimidohexanoic acid N-hydroxysuccinimide ester) ≥95%

Supplier: Matrix Scientific

076290-1G 076290-5G
102545-418EA 135.38 USD
102545-418 102545-420
EMCS (6-Maleimidohexanoic acid N-hydroxysuccinimide ester) ≥95%
EMCS (6-Maleimidohexanoic acid N-hydroxysuccinimide ester)

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₆
Melting Pt: 69…74 °C
MDL Number: MFCD00043043
CAS Number: 55750-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR