2-Bromo-4H-pyrrolo[3,4-d]thiazol-6(5H)-one ≥95%

Supplier: Matrix Scientific

076286-250MG 076286-1G 076286-5G
102545-396EA 914.25 USD
102545-396 102545-398 102545-400
2-Bromo-4H-pyrrolo[3,4-d]thiazol-6(5H)-one ≥95%
2-Bromo-4H-pyrrolo[3,4-d]thiazol-6(5H)-one

Hazard: Irritant

Formula: C₅H₃BrN₂OS
MDL Number: MFCD14706759
CAS Number: 1211531-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR