1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Phenylpyrazole-4-boronic acid, pinacol ester, 1-Phenyl-1H-pyrazole-4-boronic acid pinacol ester

076283-250MG 076283-1G 076283-5G
102545-378EA 958.46 USD
102545-378 102545-380 102545-382
1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥95%
1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₅H₁₉BN₂O₂
MDL Number: MFCD12400941
CAS Number: 1002334-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR