tert-Butyl-2-aminopropylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-aminopropylcarbamate

076282-250MG 076282-1G 076282-5G
102545-372EA 281.52 USD
102545-372 102545-374 102545-376
tert-Butyl-2-aminopropylcarbamate ≥95%
tert-Butyl-2-aminopropylcarbamate

Hazard: Irritant

Formula: C₈H₁₈N₂O₂
MDL Number: MFCD09788609
CAS Number: 255735-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR