2-Amino-3-chlorophenol ≥95%
Supplier: Matrix Scientific
076281-250MG
076281-1G
076281-5G
102545-366EA
280
USD
102545-366
102545-368
102545-370
2-Amino-3-chlorophenol ≥95%
2-Amino-3-chlorophenol
Hazard: Irritant
Formula:
C₆H₆ClNO |
MDL Number:
MFCD11848418 CAS Number: 56962-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...