(S)-1-(Pyridin-2-yl)ethanamine ≥95%

Supplier: Matrix Scientific

Synonyms: (-)-2-[(1S)-1-Aminoethyl]pyridine

076280-250MG 076280-1G 076280-5G
102545-360EA 1126.15 USD
102545-360 102545-362 102545-364
(S)-1-(Pyridin-2-yl)ethanamine ≥95%
(S)-1-(Pyridin-2-yl)ethanamine

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.1700 g/mol
MDL Number: MFCD08752493
CAS Number: 27854-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR