5-Trifluoromethyl-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%

Supplier: Matrix Scientific

076279-250MG 076279-1G
102545-356EA 684.62 USD
102545-356 102545-358
5-Trifluoromethyl-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%
5-Trifluoromethyl-1,2,3,4-tetrahydroquinoline hydrochloride

Hazard: Irritant

Formula: C10H11ClF3N
CAS Number: 1260683-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR