2-Amino-6-bromoquinoline ≥95%
Supplier: Matrix Scientific
076278-250MG
076278-1G
102545-352EA
826.15
USD
102545-352
102545-354
2-Amino-6-bromoquinoline ≥95%
2-Amino-6-bromoquinoline
Hazard: Irritant
Formula:
C₉H₇BrN₂ |
MDL Number:
MFCD06738670 CAS Number: 791626-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...