2-Methoxy-6-methylbenzoic acid ≥95%

Supplier: Matrix Scientific

076275-1G 076275-5G
102545-340EA 56.92 USD
102545-340 102545-342
2-Methoxy-6-methylbenzoic acid ≥95%
2-Methoxy-6-methylbenzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₃
MDL Number: MFCD00463871
CAS Number: 6161-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR