4-(2-Chloroethyl)morpholine ≥95%

Supplier: Matrix Scientific

076268-1G 076268-5G 076268-25G
102545-304EA 41.54 USD
102545-304 102545-306 102552-026
4-(2-Chloroethyl)morpholine ≥95%
4-(2-Chloroethyl)morpholine

Hazard: Irritant/Stored under Argon

Formula: C₆H₁₂ClNO
MW: 149.6186 g/mol
Density: 1.11
MDL Number: MFCD00603731
CAS Number: 3240-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR