1H-Pyrazolo[3,4-c]pyridine ≥95%

Supplier: Matrix Scientific

076266-1G 076266-5G
102545-294EA 123.08 USD
102545-294 102545-296
1H-Pyrazolo[3,4-c]pyridine ≥95%
1H-Pyrazolo[3,4-c]pyridine

Hazard: Irritant

Formula: C₆H₅N₃
MW: 119.1240 g/mol
MDL Number: MFCD00661308
CAS Number: 271-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR