2,6-Dichloro-3-fluorobenzoic acid ≥95%

Supplier: Matrix Scientific

076264-1G 076264-5G 076264-10G
102545-280EA 355.38 USD
102545-280 102545-282 102545-284
2,6-Dichloro-3-fluorobenzoic acid ≥95%
2,6-Dichloro-3-fluorobenzoic acid

Hazard: Irritant

Formula: C₇H₃Cl₂FO₂
MDL Number: MFCD06658165
CAS Number: 178813-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR