4-Boc-(3-hydroxymethyl)morpholine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Boc-(3-Hydroxymethyl)morpholine

076262-1G 076262-5G 076262-10G
102545-268EA 889.23 USD
102545-268 102545-270 102545-272
4-Boc-(3-hydroxymethyl)morpholine ≥95%
4-Boc-(3-hydroxymethyl)morpholine

Hazard: Irritant

Formula: C₁₀H₁₉NO₄
MW: 217.2622 g/mol
MDL Number: MFCD04038649
CAS Number: 473923-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR