5-Bromo-6-methylpyrazin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-bromo-6-methylpyrazine

076260-250MG 076260-1G 076260-5G
102545-258EA 640 USD
102545-258 102545-260 102545-262
5-Bromo-6-methylpyrazin-2-amine ≥95%
5-Bromo-6-methylpyrazin-2-amine

Hazard: Irritant

Formula: C₅H₆BrN₃
MW: 188.0300 g/mol
MDL Number: MFCD08705763
CAS Number: 74290-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR