5-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine ≥95%

Supplier: Matrix Scientific

076259-250MG 076259-1G 076259-5G
102545-252EA 400 USD
102545-252 102545-254 102545-256
5-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine ≥95%
5-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine

Hazard: Irritant

Formula: C₆H₅BrN₄
MW: 213.0400 g/mol
MDL Number: MFCD11655928
CAS Number: 1010120-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR