5-Bromoquinoline-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
076258-250MG
076258-1G
102545-248EA
1209.23
USD
102545-248
102545-250
5-Bromoquinoline-2-carboxylic acid ≥95%
5-Bromoquinoline-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₆BrNO₂ MW: 252.0641 g/mol |
MDL Number:
MFCD09881345 CAS Number: 1017412-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...