3-Boc-amino-4-bromopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Boc-Amino-4-bromopyridine

076256-1G 076256-5G
102545-240EA 710.77 USD
102545-240 102545-242
3-Boc-amino-4-bromopyridine ≥95%
3-Boc-amino-4-bromopyridine

Hazard: Irritant

Formula: C10H13BrN2O2
MW: 273.1300 g/mol
MDL Number: MFCD05664029
CAS Number: 885275-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR