Ethyl-3-amino-7-bromoquinoline-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-amino-7-bromoquinoline-2-carboxylate

076255-250MG 076255-1G
102545-236EA 967.69 USD
102545-236 102545-238
Ethyl-3-amino-7-bromoquinoline-2-carboxylate ≥95%
Ethyl-3-amino-7-bromoquinoline-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁BrN₂O₂
MDL Number: MFCD13195320
CAS Number: 1260807-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR