6-Chloro-5-nitronicotinic acid ≥95%
Supplier: Matrix Scientific
076251-1G
076251-5G
102545-216EA
61.54
USD
102545-216
102545-218
6-Chloro-5-nitronicotinic acid ≥95%
6-Chloro-5-nitronicotinic acid
Hazard: Irritant
Formula:
C₆H₃ClN₂O₄ MW: 202.5520 g/mol Boiling Pt: 401°C Melting Pt: 99-102°C Density: 1.702 Flash Pt: 196°C |
MDL Number:
MFCD09027297 CAS Number: 7477-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...