2-Methyl-4-pyrimidinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methylpyrimidin-4-amine

076250-1G 076250-5G
102545-212EA 341.54 USD
102545-212 102545-214
2-Methyl-4-pyrimidinamine ≥95%
2-Methyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₅H₇N₃
MW: 109.1292 g/mol
MDL Number: MFCD00626295
CAS Number: 74-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR