Methyl-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate

076247-250MG 076247-1G 076247-5G
102545-196EA 498.46 USD
102545-196 102545-198 102545-200
Methyl-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate ≥95%
Methyl-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MW: 193.2000 g/mol
MDL Number: MFCD08544342
CAS Number: 758684-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR