5-Fluoro-2-iodopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Iodo-5-fluoropyridine

076243-250MG 076243-1G 076243-5G
102545-172EA 456.92 USD
102545-172 102545-174 102545-176
5-Fluoro-2-iodopyridine ≥95%
5-Fluoro-2-iodopyridine

Hazard: Irritant

Formula: C5H3FIN
MW: 222.99 g/mol
MDL Number: MFCD07787574
CAS Number: 159870-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR