3-Bromopyrazolo[1,5-a]pyridine ≥95%

Supplier: Matrix Scientific

076242-250MG 076242-1G 076242-5G
102545-166EA 456.92 USD
102545-166 102545-168 102545-170
3-Bromopyrazolo[1,5-a]pyridine ≥95%
5-Bromopyrazolo[1,5-a]pyridine

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Melting Pt: 55°C
MDL Number: MFCD06661764
CAS Number: 5910-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR