2-(Methylthio)-4-pyrimidinecarbonitrile ≥95%

Supplier: Matrix Scientific

076241-250MG 076241-1G
102545-162EA 826.15 USD
102545-162 102545-164
2-(Methylthio)-4-pyrimidinecarbonitrile ≥95%
2-(Methylthio)-4-pyrimidinecarbonitrile

Hazard: Irritant

Formula: C₆H₅N₃S
CAS Number: 1124-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR