4-Chloro-1-methyl-3-nitro-1H-quinolin-2-one ≥95%

Supplier: Matrix Scientific

076240-1G 076240-5G 076240-10G
102545-156EA 710.77 USD
102545-156 102545-158 102545-160
4-Chloro-1-methyl-3-nitro-1H-quinolin-2-one ≥95%
4-Chloro-1-methyl-3-nitro-1H-quinolin-2-one

Hazard: Irritant

Formula: C₁₀H₇ClN₂O₃
CAS Number: 79966-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR