7-tert-Butyl-2-ethyl-5,6-dihydroimidazo[1,2-a]pyrazine-2,7(8H)-dicarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
7-tert-Butyl 2-ethyl 5,6-dihydroimidazo[1,2-a]pyrazine-2,7(8H)-dicarboxylate
076238-250MG
076238-1G
076238-5G
102545-144EA
541.54
USD
102545-144
102545-146
102545-148
7-tert-Butyl-2-ethyl-5,6-dihydroimidazo[1,2-a]pyrazine-2,7(8H)-dicarboxylate ≥95%
7-tert-Butyl-2-ethyl-5,6-dihydroimidazo[1,2-a]pyrazine-2,7(8H)-dicarboxylate
Hazard: Irritant
Formula:
C₁₄H₂₁N₃O₄ MW: 295.34 g/mol |
MDL Number:
MFCD10566312 CAS Number: 1053656-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...