4-Bromo-6-hydroxypyrimidine ≥95%

Supplier: Matrix Scientific

076235-250MG 076235-1G 076235-5G
102545-128EA 423.69 USD
102545-128 102545-130 102545-132
4-Bromo-6-hydroxypyrimidine ≥95%
4-Bromo-6-hydroxypyrimidine

Hazard: Irritant

Formula: C₄H₃BrN₂O
MW: 174.9834 g/mol
MDL Number: MFCD11223276
CAS Number: 1086382-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR