3-Amino-2-bromo-6-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-6-methylpyridin-3-amine, 3-Amino-2-bromo-6-picoline, 5-Amino-6-bromo-2-picoline

076230-1G 076230-5G
102545-102EA 143.08 USD
102545-102 102545-104
3-Amino-2-bromo-6-methylpyridine ≥95%
3-Amino-2-bromo-6-methylpyridine

Hazard: Irritant

Formula: C₆H₇BrN₂
MW: 187.04
MDL Number: MFCD03095078
CAS Number: 126325-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR