4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Amino-1-oxy-2,2,6,6-tetramethylpiperidine
076224-5G
076224-25G
102545-072EA
126.15
USD
102545-072
102833-916
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) ≥95%
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO)
Hazard: Irritant/Store in Freezer
Formula:
C₉H₁₉N₂O MW: 171.26 g/mol Melting Pt: 33-39°C |
MDL Number:
MFCD00006479 CAS Number: 14691-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...